Learn More
Applied Biosystems™ TaqMan™ Mutation Detection Assays
Product Code. 15205624 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
150 μL
Unit Size:
10 units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15205624 150 μL 10 units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15205624 Supplier Applied Biosystems™ Supplier No. 4465804
Follow the Workflow - Explore the Savings
Use promo code 27967
View all promotions
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
TaqMan Mutation Detection Assays
TaqMan™ Mutation Detection Assays detect somatic mutations in genes that are associated with cancer. These assays are powered by competitive allele-specific TaqMan™ PCR (castPCR™) technology. Specific and sensitive, castPCR™ can detect rare mutated DNA among wild-type DNA.
TRUSTED_SUSTAINABILITY
Specifications
Description TaqMan™ Mutation Detection Assays, 150μL, Designed to Address these Limitations, as well as to Provide a Method for Accurately Quantifying the Percentage of Specific Cancer Mutations within a Sample
Product Type TaqMan™ Mutation Detection Assays
Quantity 150 μL