Learn More
Applied Biosystems™ TaqMan™ siRNA Assay
Product Code. p-7076288 Shop All Applied Biosystems Products
Click to view available options
No. of Reactions:
25 Reactions RT primer/75 Reactions TaqMan assay
2900 Reactions
50 Reactions RT primer/150 Reactions TaqMan assay
750 Reactions
Unit Size:
Each
Product Code. 15561074
Brand: Applied Biosystems™ 4440877
TaqMan™ siRNA Assay
TaqMan™ siRNA Assays, Made-to-Order
TRUSTED_SUSTAINABILITY
Specifications
Concentration 20X (Assay), 5X (RT Primer)
Description TaqMan™ siRNA Assay, 1 x 75μL RT primer/1 x 75μL TaqMan™ Assay, 25 rxns RT Primer/75 rxns TaqMan™ Assay, Extra Small, 5X RT primer/20X TaqMan™ Assay Concentration, Store at –15 to –25°C
Product Type TaqMan™ siRNA Assay
Quantity 1 Each
No. of Reactions 25 Reactions RT primer/75 Reactions TaqMan assay
Volume 1 x 75μL RT primer/1 x 75μL TaqMan™ assay