Learn More
TARS Mouse, anti-Human, Unconjugated, Clone: 1A11F10, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67828-1-IG-20UL
Description
TARS (Threonyl-tRNA synthetase), also known as threonine-tRNA ligase or ThrRS, belongs to the aminoacyl-tRNA synthetase family. Aminoacyl-tRNA synthetase (aaRS) catalyzes the specific esterification of an amino acid with the 3′-terminal hydroxyl group of its cognate tRNA. Specificity of aminoacylation catalyzed by aaRSs ensures the accurate translation of the genetic code in the first step of protein synthesis (PMID: 26324710). TARS has some isoforms with the molecular mass of 83, 87 kDa and other MW.
Specifications
| TARS | |
| Monoclonal | |
| 500 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat |
| Western Blot, ELISA | |
| 1A11F10 | |
| Unconjugated | |
| 20 μL | |
| 6897 | |
| Liquid |