Learn More
TARS Mouse, anti-Human, Unconjugated, Clone: 1A11F10, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67828-1-IG-20UL
 View more versions of this product
Product Code. 17291476
  • € 145.00 / 20µL
Description
TARS (Threonyl-tRNA synthetase), also known as threonine-tRNA ligase or ThrRS, belongs to the aminoacyl-tRNA synthetase family. Aminoacyl-tRNA synthetase (aaRS) catalyzes the specific esterification of an amino acid with the 3′-terminal hydroxyl group of its cognate tRNA. Specificity of aminoacylation catalyzed by aaRSs ensures the accurate translation of the genetic code in the first step of protein synthesis (PMID: 26324710). TARS has some isoforms with the molecular mass of 83, 87 kDa and other MW.
Specifications
Specifications
TARS
Monoclonal
500 μg/mL
Mouse
Primary
Human, Mouse, Rat
Western Blot, ELISA
1A11F10
Unconjugated
20 μL
6897