Learn More
TARS Mouse, anti-Human, Unconjugated, Clone: 1A11F10, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | TARS |
|---|---|
| Clone | 1A11F10 |
| Concentration | 500 μg/mL |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17291476
|
Proteintech
67828-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17281476
|
Proteintech
67828-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TARS (Threonyl-tRNA synthetase), also known as threonine-tRNA ligase or ThrRS, belongs to the aminoacyl-tRNA synthetase family. Aminoacyl-tRNA synthetase (aaRS) catalyzes the specific esterification of an amino acid with the 3′-terminal hydroxyl group of its cognate tRNA. Specificity of aminoacylation catalyzed by aaRSs ensures the accurate translation of the genetic code in the first step of protein synthesis (PMID: 26324710). TARS has some isoforms with the molecular mass of 83, 87 kDa and other MW.
Specifications
| TARS | |
| 500 μg/mL | |
| Monoclonal | |
| Liquid | |
| Human, Mouse, Rat | |
| Primary |
| 1A11F10 | |
| Western Blot, ELISA | |
| Unconjugated | |
| Mouse | |
| 6897 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.