missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R10 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94569-0.1ml
This item is not returnable.
View return policy
Description
TAS2R10 Polyclonal antibody specifically detects TAS2R10 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| TAS2R10 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| MGC126811, T2R10, T2R10 MGC126813, Taste receptor family B member 2, taste receptor, family B, member 2, taste receptor, type 2, member 10, TRB2taste receptor type 2 member 10 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TAS2R10 (NP_076410.1). TSLSIFYFLKIANFSNYIFLWLKSRTNMVLPFMIVFLLISSLLNFAYIAKILNDYKTKNDTVWDLNMYKSEYFIKQILLNLGVIFFFTLSLITCIFLIISL | |
| 0.1 mL | |
| GPCR, Protein Kinase | |
| 50839 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction