missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TAS2R10 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 470.00
Specifications
| Antigen | TAS2R10 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18686701
|
Novus Biologicals
NBP2-94569-0.02ml |
0.02 mL |
€ 188.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18689670
|
Novus Biologicals
NBP2-94569-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TAS2R10 Polyclonal antibody specifically detects TAS2R10 in Mouse, Rat samples. It is validated for Western BlotSpecifications
| TAS2R10 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| GPCR, Protein Kinase | |
| PBS (pH 7.3), 50% glycerol | |
| 50839 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| MGC126811, T2R10, T2R10 MGC126813, Taste receptor family B member 2, taste receptor, family B, member 2, taste receptor, type 2, member 10, TRB2taste receptor type 2 member 10 | |
| A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TAS2R10 (NP_076410.1). TSLSIFYFLKIANFSNYIFLWLKSRTNMVLPFMIVFLLISSLLNFAYIAKILNDYKTKNDTVWDLNMYKSEYFIKQILLNLGVIFFFTLSLITCIFLIISL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title