missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau Antibody (5B10), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50051
This item is not returnable.
View return policy
Description
Tau Monoclonal antibody specifically detects Tau in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Tau | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000 | |
| P10636, P10636 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 5B10 | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau | |
| Recombinant full length 441 amino acid version of the shortest human tau isoform. Epitope located in the peptide HVPGGGNKKIETHKLTFREN, amino acids 362-381. [UniProt# P10636] | |
| 0.1 mL | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Stem Cell Markers | |
| 4137 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction