missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau Antibody (5B10), Novus Biologicals™
Mouse Monoclonal Antibody
€ 186.00 - € 502.00
Specifications
| Antigen | Tau |
|---|---|
| Clone | 5B10 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18620517
|
Novus Biologicals
NBP2-50051 |
0.1 mL |
€ 502.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18637498
|
Novus Biologicals
NBP2-50051SS |
0.025 mL |
€ 186.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tau Monoclonal antibody specifically detects Tau in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Tau | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Alzheimers Research, Apoptosis, Cell Biology, Cellular Markers, Chaperone Mediated Autophagy (CMA), Cytokine Research, MAP Kinase Signaling, Neurodegeneration, Neuroscience, Stem Cell Markers | |
| 50% PBS, 50% glycerol | |
| DDPAC, FLJ31424, FTDP-17, G protein beta1/gamma2 subunit-interacting factor 1, MGC138549, microtubule-associated protein tau, MSTDMAPTL, MTBT1Neurofibrillary tangle protein, MTBT2, PHF-tau, PPND, tau, TAUPaired helical filament-tau | |
| Recombinant full length 441 amino acid version of the shortest human tau isoform. Epitope located in the peptide HVPGGGNKKIETHKLTFREN, amino acids 362-381. [UniProt# P10636] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 5B10 | |
| Western Blot 1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| P10636, P10636 | |
| 4137 | |
| IgG1 | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title