missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau Oligomeric Complex Antibody (TOC1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20163-0.2mg
This item is not returnable.
View return policy
Description
Tau Oligomeric Complex Monoclonal antibody specifically detects Tau Oligomeric Complex in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Tau Oligomeric Complex | |
| Monoclonal | |
| 1 mg/mL | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| TOC1 | |
| Unconjugated | |
| Tau Oligomeric Complex 1, TOC1, TOC-1 | |
| The original antibody was generated by immunizing a mouse with electro eluted cross-linked Tau dimers. | |
| 0.2 mg | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction