missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tau Oligomeric Complex Antibody (TOC1) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 267.00 - € 640.00
Specifications
| Antigen | Tau Oligomeric Complex |
|---|---|
| Clone | TOC1 |
| Concentration | 1 mg/mL |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18632904
|
Novus Biologicals
NBP3-20163-0.025mg |
0.025 mg |
€ 267.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18670345
|
Novus Biologicals
NBP3-20163-0.2mg |
0.2 mg |
€ 640.00
0.20mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Tau Oligomeric Complex Monoclonal antibody specifically detects Tau Oligomeric Complex in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Tau Oligomeric Complex | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| The original antibody was generated by immunizing a mouse with electro eluted cross-linked Tau dimers. | |
| Primary | |
| Store at 4°C for up to 3 months. For longer storage, aliquot and store at -20°C. |
| TOC1 | |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Human | |
| Tau Oligomeric Complex 1, TOC1, TOC-1 | |
| IgG2b κ | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title