missing translation for 'onlineSavingsMsg'
Learn More

TBL1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-35499-20ul

 View more versions of this product

Product Code. 30228197

  • € 190.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TBL1 Polyclonal antibody specifically detects TBL1 in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TBL1
Polyclonal
Western Blot 1:500 - 1:2000, ELISA
EBI, SMAP55, TBL1transducin (beta)-like 1, transducin (beta)-like 1X-linked, X-linked
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human TBL1 (NP_005638.1).,, Sequence:, LISILQKGLQYVEAEISINEDGTVFDGRPIESLSLIDAVMPDVVQTRQQAFREKLAQQQASAAAAAAAATAAATAATTTSAGVSHQNPSKNREATVNGEEN
20 μL
Wnt Signaling Pathway
6907
Store at -20°C. Avoid freeze-thaw cycles.
IgG
ELISA, Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.