missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF7L1/TCF3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98907-100ul
This item is not returnable.
View return policy
Description
TCF7L1/TCF3 Polyclonal antibody specifically detects TCF7L1/TCF3 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| TCF7L1/TCF3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunoprecipitation 4-8 μL/mg of lysate | |
| HMG box transcription factor 3, TCF3, TCF-3, transcription factor 7-like 1, transcription factor 7-like 1 (T-cell specific, HMG-box) | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human TCF7L1/TCF3. | |
| 100 μg | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Wnt Signaling Pathway | |
| 83439 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction