missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF7L1/TCF3 Antibody (SR2086), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-21655-100ul
This item is not returnable.
View return policy
Description
TCF7L1/TCF3 Monoclonal antibody specifically detects TCF7L1/TCF3 in Human samples. It is validated for Western Blot
Specifications
| TCF7L1/TCF3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR2086 | |
| Western Blot 1:500-1:2000 | |
| HMG box transcription factor 3, TCF3, TCF-3, transcription factor 7-like 1, transcription factor 7-like 1 (T-cell specific, HMG-box) | |
| A synthesized peptide derived from human TCF7L1/TCF3 (Uniprot #: Q9HCS4) | |
| 100 μg | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Wnt Signaling Pathway | |
| 83439 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction