missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCF7L1/TCF3 Antibody (SR2086), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | TCF7L1/TCF3 |
|---|---|
| Clone | SR2086 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18618854
|
Novus Biologicals
NBP3-21655-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613924
|
Novus Biologicals
NBP3-21655-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TCF7L1/TCF3 Monoclonal antibody specifically detects TCF7L1/TCF3 in Human samples. It is validated for Western BlotSpecifications
| TCF7L1/TCF3 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| HMG box transcription factor 3, TCF3, TCF-3, transcription factor 7-like 1, transcription factor 7-like 1 (T-cell specific, HMG-box) | |
| A synthesized peptide derived from human TCF7L1/TCF3 (Uniprot #: Q9HCS4) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR2086 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Core ESC Like Genes, Signal Transduction, Stem Cell Markers, Wnt Signaling Pathway | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 83439 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title