missing translation for 'onlineSavingsMsg'
Learn More

TCP11L2 Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-93924-0.02ml

 View more versions of this product

Product Code. 18697041

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

This item is not returnable. View return policy

Description

Description

TCP11L2 Polyclonal antibody specifically detects TCP11L2 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TCP11L2
Polyclonal
Western Blot 1:500-1:2000
MGC40368, t-complex 11 (mouse) like 2, t-complex 11 (mouse)-like 2, T-complex protein 11-like protein 2
Recombinant fusion protein containing a sequence corresponding to amino acids 1-340 of human TCP11L2 (NP_689985.1). MPFNGEKQCVGEDQPSDSDSSRFSESMASLSDYECSRQSFASDSSSKSSSPASTSPPRVVTFDEVMATARNLSNLTLAHEIAVNENFQLKQEALPEKSLAGRVKHIVHQAFWDVLDSELNADPPEFEHAIKLFEEIREILLSFLTPGGNRLRNQICEVLDTDLIRQQAEHSAVDIQGLANYVISTMGKLCAPVRDNDIRELKATGNIVEVLRQIFHVLDLMQMDMANFTIMSLRPHLQRQLVEYERTKFQEILEETPSALDQTTEWIKESVNEELFSLSESALTPGAENTSKPSLSPTLVLNNSYLKLLQWDYQKKELPETLMTDGARLQELTEKLNQLK
0.02 mL
Cell Biology
255394
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.