missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TCP11L2 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 468.00
Specifications
| Antigen | TCP11L2 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
TCP11L2 Polyclonal antibody specifically detects TCP11L2 in Human samples. It is validated for Western BlotSpecifications
| TCP11L2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 255394 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| MGC40368, t-complex 11 (mouse) like 2, t-complex 11 (mouse)-like 2, T-complex protein 11-like protein 2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-340 of human TCP11L2 (NP_689985.1). MPFNGEKQCVGEDQPSDSDSSRFSESMASLSDYECSRQSFASDSSSKSSSPASTSPPRVVTFDEVMATARNLSNLTLAHEIAVNENFQLKQEALPEKSLAGRVKHIVHQAFWDVLDSELNADPPEFEHAIKLFEEIREILLSFLTPGGNRLRNQICEVLDTDLIRQQAEHSAVDIQGLANYVISTMGKLCAPVRDNDIRELKATGNIVEVLRQIFHVLDLMQMDMANFTIMSLRPHLQRQLVEYERTKFQEILEETPSALDQTTEWIKESVNEELFSLSESALTPGAENTSKPSLSPTLVLNNSYLKLLQWDYQKKELPETLMTDGARLQELTEKLNQLK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title