Learn More
TCR alpha Antibody (F1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-22493-50ug
This item is not returnable.
View return policy
Description
TCR alpha Monoclonal specifically detects TCR alpha in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications
| TCR alpha | |
| Monoclonal | |
| 0.15 mg/mL | |
| Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10 - 1:50, Immunohistochemistry-Frozen 1:10 - 1:2000 | |
| C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody reacts with Human and Primate. | |
| Store at 4°C. Do not freeze. | |
| IgG2a |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| F1 | |
| Unconjugated | |
| P01848 | |
| TRA | |
| Human TCR V alpha-2 | |
| 50 μg | |
| Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases | |
| 6955 | |
| Human, Primate | |
| Ascites |