missing translation for 'onlineSavingsMsg'
Learn More

TCR alpha Antibody (F1), Novus Biologicals™

Mouse Monoclonal Antibody

€ 189.00

Specifications

Antigen TCR alpha
Clone F1
Concentration 0.15 mg/mL
Dilution Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10 - 1:50, Immunohistochemistry-Frozen 1:10 - 1:2000
Applications Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18654263
View Documents
Novus Biologicals
NBP2-22493-50ug
50 μg
€ 189.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TCR alpha Monoclonal specifically detects TCR alpha in Human, Primate samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen.
Specifications

Specifications

TCR alpha
0.15 mg/mL
Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Unconjugated
Mouse
Adaptive Immunity, Cell Biology, Immunology, Protein Kinase, Signal Transduction, Tyrosine Kinases
P01848
6955
Human TCR V alpha-2
Primary
Store at 4°C. Do not freeze.
F1
Flow Cytometry 1 - 2 μg, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10 - 1:50, Immunohistochemistry-Frozen 1:10 - 1:2000
Monoclonal
Ascites
RUO
Human, Primate
C), D, FLJ22602, J, MGC117436, MGC22624, MGC23964, MGC71411, T cell receptor alpha locus, T-cell antigen receptor, alpha polypeptide, T-cell receptor, alpha (V, TCRA, TCRD, TRA
TRA
IgG2a
Protein A purified
This antibody reacts with Human and Primate.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.