missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdT Antibody (4B10C3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61898-0.025ml
This item is not returnable.
View return policy
Description
TdT Monoclonal specifically detects TdT in Human samples. It is validated for Western Blot, ELISA.
Specifications
| TdT | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase | |
| Mouse | |
| 58.5 kDa | |
| 0.025 mL | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 |
| Western Blot, Flow Cytometry, ELISA, Immunohistochemistry | |
| 4B10C3 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| DNTT | |
| Purified recombinant fragment of human DNTT (AA: 52-192) expressed in E. Coli. | |
| Protein G purified | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology | |
| 1791 | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction