missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TdT Antibody (4B10C3) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 203.00 - € 552.00
Specifications
| Antigen | TdT |
|---|---|
| Clone | 4B10C3 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:500-1:2000, ELISA 1:10000 |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18227533
|
Novus Biologicals
NBP2-61898 |
0.1 mL |
€ 552.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18635168
|
Novus Biologicals
NBP2-61898-0.025ml |
0.025 mL |
€ 203.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TdT Monoclonal specifically detects TdT in Human samples. It is validated for Western Blot, ELISA.Specifications
| TdT | |
| 1 mg/mL | |
| Monoclonal | |
| Purified | |
| Adaptive Immunity, Cell Cycle and Replication, Immunology | |
| 1791 | |
| Purified recombinant fragment of human DNTT (AA: 52-192) expressed in E. Coli. | |
| Primary | |
| 58.5 kDa |
| 4B10C3 | |
| Western Blot 1:500-1:2000, ELISA 1:10000 | |
| Unconjugated | |
| Mouse | |
| deoxynucleotidyltransferase, terminal, EC 2.7.7, EC 2.7.7.31, nucleosidetriphosphate:DNA deoxynucleotidylexotransferase, TDTDNA nucleotidylexotransferase, Terminal addition enzyme, Terminal deoxynucleotidyltransferase, terminal deoxyribonucleotidyltransferase, Terminal transferase | |
| DNTT | |
| IgG1 | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title