missing translation for 'onlineSavingsMsg'
Learn More

Techne™ PrimePRO QPCR RNA detection Kit, Rotavirus A (with Mastermix)

Product Code. 15371228
Click to view available options
:
150 Reactions
This item is not returnable. View return policy

Product Code. 15371228

Brand: Techne™ TKIT11031M

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
  • Kits packed in a convenient foil pouch
  • Lyophilized (freeze dried)
  • No dry ice shipping required
  • Reduced shipping costs
  • Simple protocol, identical for each kit
  • 150 reactions per kit
  • Low cost per test
  • qPCR can be days faster, more accurate and more quantifiable than traditional microbiological methods
  • Suitable for any qPCR instrument with FAM/VIC detection

Specifications

Content And Storage Lyophilized primer and probe mix, Lyophilized master mix, Lyophilized master mix resuspension buffer, Lyophilized ROX Ampule, High-copy DNA for standard curve, Internal extraction control DNA, Beta-actin endogenous control, RNAse/DNAse free water, Reverse transcriptase (RNA kits only)
Store at ambient temperature, stable for 18 months
Detection Method qPCR
For Use With (Equipment) any qPCR instrument with FAM/VIC detection
Kit Type QPCR RNA Detection Kit
No. of Tests 150
Quantity 150 Reactions
For Use With (Application) Microbial Detection
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.