missing translation for 'onlineSavingsMsg'
Learn More

Mapa™ Temp-Tec 332 Gloves

Product Code. 15689205
Click to view available options
Size:
8
9
10
Unit Size:
6 pairs
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Product Code. 15689205

Brand: Mapa™ 34332029_CLEAN

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

Mapa™ Temp-Tec 332 Gloves combine effective thermal insulation and resistance to a wide range of chemicals including acids and aliphatic solvents.

  • Multi-purpose neoprene gloves with double-knit cotton liners
  • Broad chemical resistance against acids and aliphatic solvents
  • Excellent thermal insulation
  • Embossed pebbled finish for good grip of wet or slippery objects
  • Applications in the petrochemical and polymer industries, and chemical treatment of metals
  • Also ideal for steam cleaning, boiler room work, manufacture of adhesives, handling of moulds in the ceramic industry, handling of hot piping

Specifications

Color Black, Blue
Exterior Finish Embossed Pebbled Texture
Material Neoprene
Product Type Temp-Tec 332 Gloves
Size 10
For Use With (Application) Petrochemical and chemical industries, steam cleaning
Lining Double-knit cotton
Resistant To Aliphatic organic solvents, acids
Length (Metric) 35.5 cm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.