Learn More
tert-Butyl hydrogen succinate, 97%
Product Code. p-7055979
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 15452019
Brand: Thermo Scientific Alfa Aesar H64706.03
Request Bulk or Custom Format
CAS: 15026-17-2 | C8H14O4 | 174.20 g/mol
It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 15026-17-2
Molecular Formula C8H14O4
Molecular Weight (g/mol) 174.20
MDL Number MFCD00273441
InChI Key PCOCFIOYWNCGBM-UHFFFAOYSA-N
Synonym mono-tert-butyl succinate, 4-tert-butoxy-4-oxobutanoic acid, succinic acid mono-tert-butyl ester, tert-butyl hydrogen succinate, mono-t-butyl succinate, pubchem11616, succinic acid 1-tert-butyl ester, succinic acid 4-tert-butyl ester
PubChem CID 4139107
IUPAC Name 4-[(2-methylpropan-2-yl)oxy]-4-oxobutanoic acid
SMILES CC(C)(C)OC(=O)CCC(O)=O
Specifications
Melting Point 51°C to 54°C (literature)
Color Brown-Yellow
Flash Point >110°C (230°F)
Linear Formula HOOCCH2CH2CO2C(CH3)3
Quantity 1 g
Beilstein 1772169
Solubility Information Slightly soluble in water.
Formula Weight 174.2
Percent Purity 97%
Physical Form Solid
Chemical Name or Material tert-Butyl hydrogen succinate