missing translation for 'onlineSavingsMsg'
Learn More

Edmund Bühler GmbH™ Test Tube Rack

Constructed of Stainless steel. Edmund Bühler GmbH™ Test Tube Rack for use with hinged foot on universal tray or with clamping strip on rack system.

Brand:  Edmund Bühler GmbH™ 0052 056

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10450162

  • € 98.20 / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

  • Stainless steel construction
  • Dimensions: 289 x 100mm
  • Attach to hinged foot for angle variation (order separately)
  • For use with universal platform shakers
Specifications

Specifications

Test Tube Rack
Tubes (14 mm)
68
14 mm
14 mm
Metal
Stainless Steel
Non-sterile
289 x 100mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers