Learn More
Edmund Bühler GmbH™ Test Tube Rack
Constructed of Stainless steel. Edmund Bühler GmbH™ Test Tube Rack for use with hinged foot on universal tray or with clamping strip on rack system.
€ 98.20 - € 130.00
Product Code | Brand | Holds | Diameter (Metric) Opening | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Diameter (Metric) Opening | Price | Quantity & Availability | ||||
10796002
|
Edmund Bühler GmbH™
0052 201 |
Tubes (30 mm) | 30 mm |
€ 130.00
Each |
Estimated Shipment: 08-05-2024
Log in to see stock. |
||||
10450162
|
Edmund Bühler GmbH™
0052 056 |
Tubes (14 mm) | 14 mm |
€ 98.20
Each |
Estimated Shipment: 07-06-2024
Log in to see stock. |
||||
12736967
|
Edmund Bühler GmbH™
0052 057 |
Tubes (16 mm) | 16 mm |
€ 110.00
Each |
Estimated Shipment: 17-06-2024
Log in to see stock. |
||||
10073541
|
Edmund Bühler GmbH™
0052 058 |
Tubes (18 mm) | 18 mm |
€ 109.00
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
||||
Description
- Stainless steel construction
- Dimensions: 289 x 100mm
- Attach to hinged foot for angle variation (order separately)
- For use with universal platform shakers
Specifications
Metal | |
Stainless Steel | |
Non-sterile |