Learn More
Edmund Bühler GmbH™ Test Tube Rack
Constructed of Stainless steel. Edmund Bühler GmbH™ Test Tube Rack for use with hinged foot on universal tray or with clamping strip on rack system.
Brand: Edmund Bühler GmbH™ 0052 057
Additional Details : Weight : 0.50000kg
Description
- Stainless steel construction
- Dimensions: 289 x 100mm
- Attach to hinged foot for angle variation (order separately)
- For use with universal platform shakers
Specifications
Test Tube Rack | |
Tubes (16 mm) | |
44 | |
16 mm | |
16 mm |
Metal | |
Stainless Steel | |
Non-sterile | |
289 x 100mm |