missing translation for 'onlineSavingsMsg'
Learn More

Testosterone Antibody (3T16) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-50332

 View more versions of this product

Product Code. 18650357

  • € 386.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Testosterone Monoclonal antibody specifically detects Testosterone in Nonspecies specific samples. It is validated for ELISA, Radioimmune Assays
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Testosterone
Monoclonal
1 mg/mL
ELISA 1:100 - 1:2000, Radioimmunoassay 1:100 - 1:2000
17b-Hydroxyandrost-4-ene-3-one, 17b-Testosterone
Testosterone-3-(O-carboxymethyl)oxime coupled to bovine serum albumin
0.1 mL
Cellular Signaling, Developmental Biology
Non-species specific
Purified
ELISA, Radioimmune Assays (RIA)
3T16
Unconjugated
PBS
Mouse
Protein A purified
RUO
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.