missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Testosterone Antibody (3T16) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 214.00 - € 386.00
Specifications
| Antigen | Testosterone |
|---|---|
| Clone | 3T16 |
| Concentration | 1 mg/mL |
| Dilution | ELISA 1:100 - 1:2000, Radioimmunoassay 1:100 - 1:2000 |
| Applications | ELISA, Radioimmune Assays (RIA) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18671998
|
Novus Biologicals
NBP2-50332-0.025ml |
0.025 mL |
€ 214.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18650357
|
Novus Biologicals
NBP2-50332 |
0.1 mL |
€ 386.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Testosterone Monoclonal antibody specifically detects Testosterone in Nonspecies specific samples. It is validated for ELISA, Radioimmune AssaysSpecifications
| Testosterone | |
| 1 mg/mL | |
| ELISA, Radioimmune Assays (RIA) | |
| Unconjugated | |
| Mouse | |
| Cellular Signaling, Developmental Biology | |
| PBS | |
| Testosterone-3-(O-carboxymethyl)oxime coupled to bovine serum albumin | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 3T16 | |
| ELISA 1:100 - 1:2000, Radioimmunoassay 1:100 - 1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Non-species specific | |
| 17b-Hydroxyandrost-4-ene-3-one, 17b-Testosterone | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title