missing translation for 'onlineSavingsMsg'
Learn More

Tetrabromobisphenol A Dimethyl Ether, TRC

Product Code. 30421161
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30421161 2.5 g 2.5g
30421324 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30421161 Supplier Toronto Research Chemicals Supplier No. TRCT2913102.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 37853-61-5
Molecular Formula C17 H16 Br4 O2
Molecular Weight (g/mol) 571.92
Synonym Tetrabromobisphenol A-dimethyl Ether, Benzene, 1,1'-(1-methylethylidene)bis[3,5-dibromo-4-methoxy-, 1,1'-(1-Methylethylidene)bis[3,5-dibromo-4-methoxybenzene], 2,2-Bis(3,5-dibromo-4-methoxyphenyl)propane, Tetrabromobisphenol A dimethyl ether, Tetrabromobisphenol A methyl ether
IUPAC Name 1,3-dibromo-5-[2-(3,5-dibromo-4-methoxyphenyl)propan-2-yl]-2-methoxybenzene
SMILES COc1c(Br)cc(cc1Br)C(C)(C)c2cc(Br)c(OC)c(Br)c2

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C17H16Br4O2/c1-17(2,9-5-11(18)15(22-3)12(19)6-9)10-7-13(20)16(23-4)14(21)8-10/h5-8H,1-4H3
Formula Weight 567.7884
Chemical Name or Material Tetrabromobisphenol A Dimethyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.