Learn More
TFEB Mouse, anti-Human, Clone: 1C2D9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68632-1-IG-150UL
This item is not returnable.
View return policy
Description
The TFEB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TFEB. This antibody recognizes human antigen. The TFEB antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TFEB | |
| 1C2D9 | |
| AlphaTFEB, bHLHe35, TCFEB, TFEB, transcription factor EB | |
| 150 μL | |
| 7942 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG1 |