Learn More
TFEB Mouse, anti-Human, Clone: 1C2D9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68632-1-IG-150UL
 View more versions of this product
Product Code. 18049325
  • € 410.00 / 150µL
Description
The TFEB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TFEB. This antibody recognizes human antigen. The TFEB antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
TFEB
1C2D9
AlphaTFEB, bHLHe35, TCFEB, TFEB, transcription factor EB
150 μL
7942
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human
150 kDa