missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TFEB Mouse, anti-Human, Clone: 1C2D9, Proteintech
Mouse Monoclonal Antibody
€ 145.00 - € 410.00
Specifications
| Antigen | TFEB |
|---|---|
| Clone | 1C2D9 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18049325
|
Proteintech
68632-1-IG-150UL |
150 μL |
€ 410.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18059655
|
Proteintech
68632-1-IG-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The TFEB antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TFEB. This antibody recognizes human antigen. The TFEB antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| TFEB | |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| AlphaTFEB, bHLHe35, TCFEB, TFEB, transcription factor EB | |
| Primary |
| 1C2D9 | |
| Western Blot, ELISA | |
| Mouse | |
| 7942 | |
| IgG1 | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title