missing translation for 'onlineSavingsMsg'
Learn More

Ansell ActivArmr™ 78-101 Knitted Thermal Gloves

Product Code. 15222709 Shop All Ansell Products
Click to view available options
Size:
7
9
Unit Size:
12 pairs
Pack of 144
This item is not returnable. View return policy

Product Code. 15222709

Brand: Ansell 78101_9

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Knitted thermal work gloves feature special fibers that provide a thermal insulation barrier between the skin and surrounding environment.

  • Gloves dry faster than cotton to efficiently eliminate moisture from the skin
  • Special hollow-core fibres provide a thermal insulation barrier between the skin and surrounding environment
  • Automatic knit technology construction for longer, more comfortable wear
  • Seamless design eliminates any potential discomfort or skin irritation
  • Thermal insulation for cold weather protection
  • Ambidextrous to fit either hand
TRUSTED_SUSTAINABILITY

Specifications

ANSI Abrasion Rating ANSI 3
Certifications/Compliance (Europe) EN 420:2003 + A1:2009; Category III
Coating Material Uncoated
Color Blue
Exterior Finish No Texture
Hand Ambidextrous
Material Thermolite
Product Type Thermal Mechanic's Gloves
Size 9
EN388 Standard 214XA
For Use With (Application) Automotive Assembly, Food Processing
Glove Cuff Style Knit
Series 78-101
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.