missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGAR/C12orf5 Antibody (M2-P4H2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-2698-0.025mg
This item is not returnable.
View return policy
Description
TIGAR/C12orf5 Monoclonal specifically detects TIGAR/C12orf5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| TIGAR/C12orf5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| TIGAR | |
| Synthetic peptide; IINFEEGREV (amino acids 239-248) | |
| 0.025 mg | |
| Biologically Active Proteins, Lipid and Metabolism | |
| 57103 | |
| Human | |
| Purified |
| Western Blot | |
| M2-P4H2 | |
| Unconjugated | |
| chromosome 12 open reading frame 5, EC 3.1.3.46, probable fructose-2,6-bisphosphatase TIGAR, TIGARTP53-induced glycolysis and apoptosis regulator, transactivated by NS3TP2 protein | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| HLA DP (TAL 3C3). | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction