missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIGAR/C12orf5 Antibody (M2-P4H2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
€ 190.00
Specifications
| Antigen | TIGAR/C12orf5 |
|---|---|
| Clone | M2-P4H2 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 |
| Applications | Western Blot |
Description
TIGAR/C12orf5 Monoclonal specifically detects TIGAR/C12orf5 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TIGAR/C12orf5 | |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Biologically Active Proteins, Lipid and Metabolism | |
| chromosome 12 open reading frame 5, EC 3.1.3.46, probable fructose-2,6-bisphosphatase TIGAR, TIGARTP53-induced glycolysis and apoptosis regulator, transactivated by NS3TP2 protein | |
| TIGAR | |
| IgG1 κ | |
| Protein A purified | |
| HLA DP (TAL 3C3). |
| M2-P4H2 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:10 - 1:500, Immunohistochemistry-Paraffin 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| 57103 | |
| Synthetic peptide; IINFEEGREV (amino acids 239-248) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title