missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TIMP-3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 256.00 - € 517.00
Specifications
| Antigen | TIMP-3 |
|---|---|
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry 1-2ug/ml, Immunocytochemistry/Immunofluorescence 1-2ug/mlimmunohistochemistry 1-2ug/ml, Immunohistochemistry-Paraffin |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18451862
|
Novus Biologicals
NBP2-29478-20ug |
20 ug |
€ 256.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18143183
|
Novus Biologicals
NBP2-29478 |
0.1 mg |
€ 517.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TIMP-3 Polyclonal specifically detects TIMP-3 in Human, Mouse, Rat, Bovine, Canine, Equine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| TIMP-3 | |
| Flow Cytometry 0.5-1ug/million cells, Immunohistochemistry 1-2ug/ml, Immunocytochemistry/Immunofluorescence 1-2ug/mlimmunohistochemistry 1-2ug/ml, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Bovine, Canine, Equine, Sheep | |
| P35625 | |
| 7078 | |
| TIMP-3 (aa175-211) (Uniprot: P35625) | |
| Primary | |
| Store at 4C. | |
| 30 kDa |
| 0.2 mg/ml | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Apoptosis, Cancer, Hypoxia, Metastasis, Vision | |
| PBS with 0.05% BSA. with 0.05% Sodium Azide | |
| HSMRK222, K222, K222TA2, metalloproteinase inhibitor 3, MIG-5 protein, Protein MIG-5, pseudoinflammatory), SFD, TIMP metallopeptidase inhibitor 3, TIMP-3, tissue inhibitor of metalloproteinase 3 (Sorsby fundus dystrophy, Tissue inhibitor of metalloproteinases 3 | |
| TIMP3 | |
| IgG | |
| Protein A purified | |
| TIMP3 (tissue inhibitor of metalloproteinases 3), along with family members TIMP1, TIMP2, and TIMP4, are inhibitors of the matrix metalloproteinases (MMPs), a group of peptidases involved in degradation of the extracellular matrix (ECM). An imbalance between MMPs and the associated TIMPs may play a significant role in the invasive phenotype of malignant tumors. TIMP s inhibit the proteolytic invasiveness of tumor cells and normal placental trophoblast cells. TIMP-3 may be involved in regulating trophoblastic invasion of the uterus as well as in regulating remodeling of the extracellular matrix during the folding of epithelia, and in the formation, branching and expansion of epithelial tubes. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title