missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin, Residuals , ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Tin, Residuals , Ø40x15mm
Specifications
Specifications
| Chemical Name or Material | Cobalt/Selenium/Zinc/Aluminium/Arsenic/Gold/Iron/Nickel/Gallium/Tellurium/Germanium/Silver/Cadmium/Bismuth/Mercury/Indium/Copper/Antimony/Lead/Thallium/Magnesium/Chromium/Molybdenum/Sulfur/Calcium/Phosphorus/Tin |
| CAS | 7440-31-5 |
| Quantity | 1 Ea. |
| Form | Disc |
| Concentration or Composition (by Analyte or Components) | Cobalt 0.0017 %/Selenium 0.002 %/Zinc 0.0045 %/Aluminium 0.005 %/Arsenic 0.005 %/Gold 0.005 %/Iron 0.011 %/Nickel 0.0127 %/Gallium 0.019 %/Tellurium 0.02 %/Germanium 0.021 %/Silver 0.033 %/Cadmium 0.034 %/Bismuth 0.041 %/Mercury 0.046 %/Indium 0.048 %/Copper 0.062 %/Antimony 0.069 %/Lead 0.147 %/Thallium (0.00003) %/Magnesium (0.0002) %/Chromium (0.001) %/Molybdenum (0.001) %/Sulfur (0.002) %/Calcium (0.004) %/Phosphorus (0.004) %/Tin (99.5) % |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction