missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tin, Technical, Metal, Granulated, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Specifications
Specifications
| Color | Silver |
| Melting Point | 231.9°C |
| Boiling Point | 2270°C |
| Quantity | 500 g |
| Packaging | Plastic powder jar |
| Vapor Pressure | 1mmHg 1492 |
| Form | Solid |
| Formula Weight | 118.69g/mol |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction