Learn More
Titanium, Ti 6-4 ELI, Grade 23, R56407, ARMI∣MBH
Click to view available options
Quantity:
1 Ea.
Unit Size:
Each
Description
Titanium, Ti 6-4 ELI, Grade 23, R56407
Specifications
Specifications
| Chemical Name or Material | Aluminium, Boron, Carbon, Chromium, Cobalt, Copper, Hydrogen, Iron, Manganese, Molybdenum, Nickel, Niobium, Nitrogen, Oxygen, Silicon, Sulfur, Tin, Titanium, Vanadium, Yttrium, Zirconium |
| Quantity | 1 Ea. |
| Diameter (Metric) Outside | 38 mm |
| Form | Disc |
| Thickness (Metric) | 19 mm |
| Concentration or Composition (by Analyte or Components) | Cobalt: 17 ppm, Boron: 12 ppm, Yttrium: 11 ppm, Manganese: 0.0004%, Sulfur: 0.0007%, Hydrogen: 0.0012%, Copper: 0.002%, Zirconium: 0.0021%, Niobium: 0.004%, Molybdenum: 0.005%, Tin: 0.006%, Nitrogen: 0.0061%, Chromium: 0.0139%, Carbon: 0.016%, Nickel: 0.017%, Silicon: 0.017%, Oxygen: 0.119%, Iron: 0.183%, Vanadium: 3.88%, Aluminium: 6.26% |