Learn More
TLR11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56742
This item is not returnable.
View return policy
Description
TLR11 Polyclonal specifically detects TLR11 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.