missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 221.00 - € 469.00
Specifications
| Antigen | TLR11 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 1-3 ug/ml, Flow Cytometry 0.5-1 ug/10-6 cells |
| Applications | Western Blot, Flow Cytometry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18068385
|
Novus Biologicals
NB100-56742SS |
0.025 mg |
€ 221.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18745763
|
Novus Biologicals
NB100-56742 |
€ 469.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
TLR11 Polyclonal specifically detects TLR11 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.Specifications
| TLR11 | |
| Western Blot 1-3 ug/ml, Flow Cytometry 0.5-1 ug/10-6 cells | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| NP_991388 | |
| 442887 | |
| A synthetic peptide within amino acid residues 911-926 (CEQLKRRLSKAGQERD) of mouse TLR11 (Genbank accession no. NP_991388) was used as immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 1.0 mg/mL | |
| Western Blot, Flow Cytometry | |
| Unconjugated | |
| Rabbit | |
| Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors | |
| PBS with 0.05% Sodium Azide | |
| hgnc31753, Toll like receptor 11 | |
| TLR11 | |
| IgG | |
| Protein G purified | |
| Immunizing peptide will block the mouse TLR11 band in western blotting. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title