missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR11 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-56742SS
This item is not returnable.
View return policy
Description
TLR11 Polyclonal specifically detects TLR11 in Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| TLR11 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| hgnc31753, Toll like receptor 11 | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| Immunizing peptide will block the mouse TLR11 band in western blotting. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Flow Cytometry | |
| 1.0 mg/mL | |
| Western Blot 1-3 ug/ml, Flow Cytometry 0.5-1 ug/10-6 cells | |
| NP_991388 | |
| TLR11 | |
| A synthetic peptide within amino acid residues 911-926 (CEQLKRRLSKAGQERD) of mouse TLR11 (Genbank accession no. NP_991388) was used as immunogen. | |
| 0.025 mg | |
| Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors | |
| 442887 | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction