Learn More
TLR8 Monoclonal Antibody (OTI3A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA810175S
This item is not returnable.
View return policy
Description
TLR8 Monoclonal antibody specifically detects TLR8 in Human samples. It is validated for Western BlotSpecifications
| TLR8 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NR97 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 51311 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI3A6 | |
| Unconjugated | |
| TLR8 | |
| CD antigen 288; CD288; TLR8; toll like receptor 8; toll-like receptor 8; UNQ249/PRO286 | |
| Human recombinant protein fragment corresponding to amino acids 32-153 of human TLR8 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |