Learn More
TLR8 Monoclonal Antibody (OTI3A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA810175S
Encompass_Preferred
 View more versions of this product
Product Code. 17513074
  • € 242.00 / 30µL
Description
TLR8 Monoclonal antibody specifically detects TLR8 in Human samples. It is validated for Western Blot
Specifications
Specifications
TLR8
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NR97
Mouse
Affinity chromatography
RUO
51311
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI3A6
Unconjugated
TLR8
CD antigen 288; CD288; TLR8; toll like receptor 8; toll-like receptor 8; UNQ249/PRO286
Human recombinant protein fragment corresponding to amino acids 32-153 of human TLR8 produced in E.coli.
30 μL