missing translation for 'onlineSavingsMsg'
Learn More

TLR8 Monoclonal Antibody (OTI3A6), TrueMAB™, OriGene

Mouse Monoclonal Antibody

€ 242.00 - € 540.87

Specifications

Antigen TLR8
Clone OTI3A6
Concentration 1 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17513074
Encompass_Preferred
View Documents
Origene Technologies
TA810175S
30 μL
€ 242.00
30µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17593064
Encompass_Preferred
View Documents
Origene Technologies
TA810175
100 μL
€ 540.87
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TLR8 Monoclonal antibody specifically detects TLR8 in Human samples. It is validated for Western Blot
Specifications

Specifications

TLR8
1 mg/mL
Monoclonal
Liquid
RUO
Q9NR97
IgG1
-20° C, Avoid Freeze/Thaw Cycles
OTI3A6
Western Blot
Unconjugated
Mouse
Human
51311
Primary
TLR8
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.