missing translation for 'onlineSavingsMsg'
Learn More

TLR9 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 3 publications

Brand:  Novus Biologicals NBP1-76680-0.025mg

 View more versions of this product

Product Code. 18597740

  • € 189.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TLR9 Polyclonal specifically detects TLR9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TLR9
Polyclonal
Unconjugated
PBS with 0.02% Sodium Azide
CD289, CD289 antigen, toll-like receptor 9
Rabbit
97 kDa
0.025 mg
Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors
54106
Human
IgG
Western Blot
1 mg/mL
Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-20 μg/mL
AAH32713
TLR9
Antibody was raised against a peptide corresponding to 15 amino acids near the center of human TLR9. The immunogen is located within amino acids 840 - 890 of TLR9. Amino Acid Squence: RGRQSGRDEDALPYD
Affinity Purified
RUO
Primary
At least two isoforms of TLR9 are known to exist; this antibody will detect both isoforms. TLR9 antibody is predicted to not cross-react with other TLR proteins.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only