missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TLR9 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 3 publications
€ 189.00
Specifications
| Antigen | TLR9 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-20 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
TLR9 Polyclonal specifically detects TLR9 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence, Immunocytochemistry.Specifications
| TLR9 | |
| Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunocytochemistry/Immunofluorescence 2-20 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Cytokine Research, Immunology, Innate Immunity, Toll Like Receptors | |
| PBS with 0.02% Sodium Azide | |
| CD289, CD289 antigen, toll-like receptor 9 | |
| TLR9 | |
| IgG | |
| Affinity Purified | |
| At least two isoforms of TLR9 are known to exist; this antibody will detect both isoforms. TLR9 antibody is predicted to not cross-react with other TLR proteins. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| AAH32713 | |
| 54106 | |
| Antibody was raised against a peptide corresponding to 15 amino acids near the center of human TLR9. The immunogen is located within amino acids 840 - 890 of TLR9. Amino Acid Squence: RGRQSGRDEDALPYD | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| 97 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title