missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TM2D2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marque: Novus Biologicals NBP2-30462
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
TM2D2 Polyclonal specifically detects TM2D2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Spécification
| TM2D2 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q9BX73 | |
| TM2D2 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: TASQELGYGCLKFGGQAYSDVEHTSVQCHALDGIECASPRTFLRENKPCIKYTG | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| BBP-like protein 1, Beta-amyloid-binding protein-like protein 1, BLP1MGC125814, MGC125813, TM2 domain containing 2, TM2 domain-containing protein 2 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 83877 | |
| Human | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
For Research Use Only
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu