missing translation for 'onlineSavingsMsg'
Learn More

TM2D2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 280.00 - € 589.00

Specifications

Antigen TM2D2
Dilution Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18443402
View Documents
Novus Biologicals
NBP2-30462-25ul
25ul
€ 280.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18162525
View Documents
Novus Biologicals
NBP2-30462
0.1 mL
€ 589.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TM2D2 Polyclonal specifically detects TM2D2 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications

Specifications

TM2D2
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Unconjugated
RUO
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
BBP-like protein 1, Beta-amyloid-binding protein-like protein 1, BLP1MGC125814, MGC125813, TM2 domain containing 2, TM2 domain-containing protein 2
TM2D2
IgG
Affinity Purified
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Western Blot 0.4 ug/ml, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200
Polyclonal
Rabbit
Human
Q9BX73
83877
This antibody was developed against a recombinant protein corresponding to amino acids: TASQELGYGCLKFGGQAYSDVEHTSVQCHALDGIECASPRTFLRENKPCIKYTG
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.