missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMC1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-05608-100ul
This item is not returnable.
View return policy
Description
TMC1 Polyclonal antibody specifically detects TMC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TMC1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| DFNA36, DFNB11, DFNB7, transmembrane channel-like 1, transmembrane channel-like protein 1, Transmembrane cochlear-expressed protein 1, transmembrane, cochlear expressed, 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 600-700 of human TMC1 (NP_619636.2). INILRLHTSMYFQCWAVMCCNVPEARVFKASRSNNFYLGMLLLILFLSTMPVLYMIVSLPPSFDCGPFSGKNRMFEVIGETLEHDFPSWMAKILRQLSNPG | |
| 100 μg | |
| Neuroscience | |
| 117531 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction