missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMC1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 188.00 - € 463.00
Specifications
| Antigen | TMC1 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18622068
|
Novus Biologicals
NBP3-05608-100ul |
100 μg |
€ 463.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18600987
|
Novus Biologicals
NBP3-05608-20ul |
20 μg |
€ 188.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TMC1 Polyclonal antibody specifically detects TMC1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| TMC1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Neuroscience | |
| PBS with 50% glycerol, pH7.3. | |
| 117531 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| DFNA36, DFNB11, DFNB7, transmembrane channel-like 1, transmembrane channel-like protein 1, Transmembrane cochlear-expressed protein 1, transmembrane, cochlear expressed, 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 600-700 of human TMC1 (NP_619636.2). INILRLHTSMYFQCWAVMCCNVPEARVFKASRSNNFYLGMLLLILFLSTMPVLYMIVSLPPSFDCGPFSGKNRMFEVIGETLEHDFPSWMAKILRQLSNPG | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title