missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM189 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94610-0.1ml
This item is not returnable.
View return policy
Description
TMEM189 Polyclonal antibody specifically detects TMEM189 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TMEM189 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:100, Immunohistochemistry-Paraffin | |
| KUA, transmembrane protein 189, UBE2V1, ubiquitin-conjugating enzyme E2 variant 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 171-270 of human TMEM189 (NP_954580.1). FVFCLIIFGTFTNQIHKWSHTYFGLPRWVTLLQDWHVILPRKHHRIHHVSPHETYFCITTGWLNYPLEKIGFWRRLEDLIQGLTGEKPRADDMKWAQKIK | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 387521 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction